Cyclic di-GMP (sodium salt) ≥95%

Supplier: Cayman Chemical Company

Synonyms: Cyclic diguanylate, c-di-GMP, 3',5'-Cyclic diguanylic acid, Guanylyl-(3'→5')-3'-guanylic acid, cyclic nucleotide, disodium salt

17144-500 17144-1
76293-128EA 145.01 USD
76293-128 76293-130
Cyclic di-GMP (sodium salt) ≥95%
Cyclic di-GMP (sodium salt)

  • A bacterial second messenger and cyclic dinucleotide


A bacterial second messenger and cyclic dinucleotide; an important second messenger in bacteria involved in biofilm formation, motility, virulence, and cell cycling; detected by and binds to STING in eukaryotic cells, activating the innate immune system (Kd = 1.21 µM); induces IFN-β mRNA expression in vitro (EC50 = 537.8 nM) less potently than other cyclic dinucleotides

Formula: C₂₀H₂₂N₁₀O₁₄P₂ ·2Na
MW: 734.4 g/mol
Storage Temperature: Freezer
CAS Number: 2222132-40-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14c33c2ebb9bd298314239f3923121c6945bf398&vid=ea7cfd90220c67407f9d7275c7ceed9f1e6ed700&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81f090f637703e6eaf9c1e95b3a42c49e4621fc2&vid=81f090f637703e6eaf9c1e95b3a42c49e4621fc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A solid
Purity ≥95%
Stability ≥ 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR