Cyclic di-AMP (sodium salt) ≥98%

Supplier: Cayman Chemical Company

Synonyms: 3',5'-Cyclic diadenylic acid, Cyclic diadenylate, Adenylyl-(3'→5')-3'-adenylic acid, cyclic nucleotide, disodium salt, c-di-AMP, Cyclic di-Adenosine monophosphate

17753-500 17753-1 17753-5
76293-122EA 149.28 USD
76293-122 76293-124 76293-126
Cyclic di-AMP (sodium salt) ≥98%
Cyclic di-AMP (sodium salt)

  • A bacterial second messenger


A second messenger produced by bacteria but not by mammals; impacts bacterial cell growth, cell wall homeostasis, pathogenicity, and other cellular functions; triggers the expression of interferon genes in mammalian cells

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e7693b19141ef9939324733a1b17ea8e0fe08f4&vid=727de4af11186f3666719902d088ec59e731ebf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR