G10 (Sting antagonist) ≥98%
Supplier: Cayman Chemical Company
- An indirect activator of STING signaling
An indirect activator of STING signaling; induces IRF3- and IFN-dependent transcription in human fibroblasts, PBMCs, and HUMECs; prevents replication of chikungunya and Venezuelan equine encephalitis alphaviruses (IC90s = 8.01 and 24.57 µM, respectively); increases phosphorylation of IRF3 when used at a concentration of 100 µM but not in cells lacking STING
Formula:
C₂₁H₁₆ClFN₂O₃S Storage Temperature: Freezer |
MDL Number:
MFCD04062826 CAS Number: 702662-50-8 |
|
Specification Test Results
Formulation | A crystalline solid |
Purity | ≥98% |
Stability | ≥ 2 years |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...