IKK-2 ≥98%

Supplier: Cayman Chemical Company

Synonyms: IMD-0354, N-(3,5-Bis-trifluoromethylphenyl)-5-chloro-2-hydroxybenzamide, N-(3,5-Bis(trifluoromethyl)phenyl)-5-chloro-2-hydroxybenzamide

17290-1 17290-5 17290-10 17290-50
76293-038EA 45.88 USD
76293-038 76293-040 76293-042 76293-044
IKK-2 ≥98%
IKK-2

  • An IKKβ (IKK2) inhibitor


An IKKβ (IKK2) inhibitor that blocks NF-κB phosphorylation (IC50 = ~250 nM) and subsequent NF-κB p65 nuclear translocation; exhibits cardioprotective and anti-cancer properties

Formula: C₁₅H₈ClF₆NO₂
MW: 383.67 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00218820
CAS Number: 978-62-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=882d9f6ea3311c9e2ba77c5c6f5dc3b9e8504ac0&vid=882d9f6ea3311c9e2ba77c5c6f5dc3b9e8504ac0&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b621d23b74c6db948dc73a152bf95f2852999e19&vid=b621d23b74c6db948dc73a152bf95f2852999e19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR