CAY10576 ≥95%
Supplier: Cayman Chemical Company
Synonyms:
IKK-3, 5-(5,6-dimethoxy-1H-benzimidazol-1-yl)-3-[[2-(methylsulphonyl)phenyl]methoxy]-2-thiophenecarbonitrile, CAY10576
10011249-1
10011249-5
10011249-10
10011249-50
76292-976EA
54.06
USD
76292-976
76292-978
76292-980
76292-982
CAY10576 ≥95%
IKK-ε Kinase
- A potent and selective inhibitor of IKK-ε
A benzimidazole analog that selectively inhibits IKK-ε with an IC50 value of 40 nM and is essentially inactive at IKK-α and IKK-β
Formula:
C₂₂H₁₉N₃O₅S₂ MW: 469.5 g/mol Storage Temperature: Freezer |
CAS Number:
862812-98-4 |
Specification Test Results
Formulation | A crystalline solid |
Purity | ≥95% |
Stability | ≥ 2 years |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...