TPCA-1 ≥98%
Supplier: Cayman Chemical Company
Synonyms:
5-(4-Fluorophenyl)-2-ureidothiophene-3-carboxamide, 2-[(aminocarbonyl)amino]-5-(4-fluorophenyl)-3-thiophenecarboxamide, IKK2 Inhibitor IV
15115-500
15115-1
15115-5
15115-10
76293-068EA
81.7
USD
76293-068
76293-070
76293-072
76293-074
TPCA-1 ≥98%
TPCA-1
- A selective inhibitor of IKK2
A selective inhibitor of IKK2 (IC50s = 17.9 nM and 0.40 μM for IKK2 and IKK1, respectively); reduces the severity and onset of collagen-induced arthritis in mice and blocks cytokine release in an antigen-driven rat model of lung inflammation
Formula:
C₁₂H₁₀FN₃O₂S MW: 279.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD09037541 CAS Number: 507475-17-4 |
Specification Test Results
Formulation | A crystalline solid |
Purity | ≥98% |
Stability | ≥ 2 years |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...