NF-κB Inhibitor (SN50)
Supplier: Cayman Chemical Company
17493-500 17493-1 17493-5
76293-018EA 80.38 USD
76293-018 76293-020 76293-022
NF-κB Inhibitor (SN50)
Proteins and Peptides
A synthetic peptide corresponding to the nuclear localization sequence of NF-κB p105 subunit (also known as p50) appended to a hydrophobic sequence to facilitate import into living cells; blocks the nuclear import of p105 in cells treated with activators of NF-κB signaling, including LPS and TNF-α.
  • A cell-permeable peptide that blocks nuclear import of NF-κB
Stability: ≥2 years
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14c33c2ebb9bd298314239f3923121c6945bf398&vid=ea7cfd90220c67407f9d7275c7ceed9f1e6ed700&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a85744dd71af5832d370851b8ad1f914ee9b0be&vid=9a85744dd71af5832d370851b8ad1f914ee9b0be&prid=web&clid=SAPDAM