QNZ ≥98%

Supplier: Cayman Chemical Company

Synonyms: 6-Amino-4-[2-(4-phenoxyphenyl)ethylamino]quinazoline, N4-(4-Phenoxyphenethyl)quinazoline-4,6-diamine

10006734-500 10006734-1 10006734-5 10006734-10
76293-096EA 24.8 USD
76293-096 76293-098 76293-100 76293-102
QNZ ≥98%
QNZ

  • An inhibitor of NF-κB activation


An inhibitor of NF-κB activation with an IC50 value of 11 nM in human Jurkat cells; inhibits TNF-α production from LPS-stimulated mouse splenocytes (IC50 = 7 nM)

Formula: C₂₂H₂₀N₄O
MW: 356.429 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06411436
CAS Number: 545380-34-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ad0ea0de92022ef1a590ac0a65e4dcd44ce469c&vid=4ad0ea0de92022ef1a590ac0a65e4dcd44ce469c&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e58edbe3a3465ce3c43ae51ba0ffe67f7bd040a0&vid=e58edbe3a3465ce3c43ae51ba0ffe67f7bd040a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR