Phenethyl caffeate ≥98%

Supplier: Cayman Chemical Company

Synonyms: 3,4-Dihydroxycinnamic Acid Phenethyl Ester, Caffeic Acid Phenethyl Ester, Phenethyl 3-(3,4-dihydroxyphenyl)acrylate, CAPE, Phenethyl 3,4-Dihydroxycinnamate

70750-1 70750-50 70750-100 70750-500
76292-990EA 502.44 USD
76292-990 76293-114 76293-116 76293-118
Phenethyl caffeate ≥98%
Phenethyl caffeate

  • A potent inhibitor of NF-κB activation


A potent and specific inhibitor of NF-kB that exhibits antimitogenic, anticarcinogenic, antiinflammatory, and immunomodulatory properties

Formula: C₁₇H₁₆O₄
MW: 284.31 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00866470
CAS Number: 104594-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3cf1ccb585195a32dff90ac4686a7c48b096374&vid=e3cf1ccb585195a32dff90ac4686a7c48b096374&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51b1cef2c88757f722b05106424f863ddbd6ba0c&vid=51b1cef2c88757f722b05106424f863ddbd6ba0c&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=594b3e28b036e610a45dc9fef83c43b7ac179968&vid=594b3e28b036e610a45dc9fef83c43b7ac179968&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 1 year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR