Dimethyl biphenyl-4,4'-dicarboxylate ≥95%

Supplier: Cayman Chemical Company

17869-10 17869-25 17869-50 17869-100
76292-932EA 60.78 USD
76292-932 76292-934 76292-936 76292-938
Dimethyl biphenyl-4,4'-dicarboxylate ≥95%
Dimethyl biphenyl-4,4'-dicarboxylate

  • A hepatoprotectant


A planar biphenyldicarboxylic acid ester used as a hepatoprotectant in the treatment of chronic hepatitis; 250 µg/ml stimulates JAK/STAT signaling and induces the expression of interferon-α stimulated genes in a HepG2 cell line

Formula: C₁₆H₁₄O₄
Storage Temperature: Freezer
MDL Number: MFCD00017201
CAS Number: 792-74-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=978fbb4db6478c4e789b89cbd1dfd1e3326a3f07&vid=978fbb4db6478c4e789b89cbd1dfd1e3326a3f07&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fe7ac33c06b58b077c986e1231125dc88dac4ac&vid=88176a9d46a9a7b040790a07a28cc50f9183107d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥95%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR