Inlet Thermometer Adapters, PTFE, Chemglass
Supplier: Chemglass
CG-1048-01 CG-1048-02 CG-1048-04 CG-1048-05 CG-1048-06
80060-308EA 51.27 USD
80060-308 80060-310 80060-312 80060-314 80060-316
Inlet Thermometer Adapters, PTFE, Chemglass
Adapters Adapters, Conical Ground Joints
These PTFE adapters have an inner joint of the listed size.
  • Made entirely of PTFE
The compression nut allows for vertical adjustment of thermometers or bleed tubes having an O.D. between 6 and 7 mm.
Información de suministro: Supplied complete with one #108 Viton O-Ring.