Splash Guard Adapters, Chemglass
Supplier: Chemglass
CG-1041-01 CG-1041-03 CG-1041-05
89028-210EA 117.12 USD
89028-210 89028-214 89028-218
Splash Guard Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters prevent splash-up when placed between the distilling flask and columm.
  • Comes with a choice of full or medium length joints
  • Overall height is minimal to aid in effective distillation
  • Manufactured by Chemglass
Adapter with a choice of full or medium length joints. When placed between the distilling flask and columm, they prevent splash-ups.