Adapters, Sanitary to Standard Taper, Stainless Steel, Tri-Clamp, Chemglass
Supplier: Chemglass
CG-1002-B-20 CG-1002-B-22
89492-742EA 272.03 USD
89492-742 89492-744
Adapters, Sanitary to Standard Taper, Stainless Steel, Tri-Clamp, Chemglass
Adapters Adapters, Conical Ground Joints
Provides a convenient way to connect sanitary fittings and adapters to Chemglass reaction systems.
  • Approximate overall length is 4" (102 mm)
  • Stainless steel
  • Manufactured by Chemglass
Standard taper inner joint to sanitary fitting adapter.
Precaución: NOTE: Clamp and seals must be purchased separately.