Airfree® Schlenk Addition Funnels, Graduated, with Glass T-Bore Stopcock, Chemglass
Supplier: Chemglass
AF-0547-G-01 AF-0547-G-02
80064-912EA 519.36 USD
80064-912 80064-914
Airfree® Schlenk Addition Funnels, Graduated, with Glass T-Bore Stopcock, Chemglass
Funnels Dropping Funnels
Pressure equalizing addition funnel, with graduations and a glass T-Bore stopcock in the equalizing arm to permit purging and isolation of the funnel contents for storage.
  • Glass T-Bore stopcock
  • Top and lower inner joints are the same size
  • 2 mm high vacuum glass stopcocks
Both top outer and lower inner joints are the same size. Lower stopcock is a glass straight bore stopcock. Both stopcocks are 2 mm high vacuum glass stopcocks with a stainless steel constant pressure retention device.