Single Neck Storage Bottles, Chemglass
Supplier: Chemglass
CG-1126-03 CG-1126-05 CG-1126-07 CG-1126-11 CG-1126-12 CG-1126-15 CG-1126-01
80066-380EA 127.44 USD
80066-380 80066-382 80066-384 80066-390 80066-392 80066-396 80066-376
Single Neck Storage Bottles, Chemglass
Bottles Glass Bottles
Bottles have a standard taper outer joint.
Bottles are available amberized for protection of light sensitive materials.
Información de suministro: Supplied without stopper.