Airfree® Schlenk Reaction Tubes, Single Neck, [ST] Inner Joint, 2 mm Stopcock, Chemglass
Supplier: Chemglass
AF-0538-01 AF-0538-03 AF-0538-04 AF-0538-05
80064-742EA 200.28 USD
80064-742 80064-746 80064-748 80064-750
Airfree® Schlenk Reaction Tubes, Single Neck, [ST] Inner Joint, 2 mm Stopcock, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Cylindrical shaped tube, with a standard taper inner joint, useful for simple reactions.
  • 2 mm high vacuum glass stopcock
  • Stainless steel constant pressure retaining device