Complete Capillary Injector Add-On Kits, PerkinElmer
Supplier: Revvity Health Sciences
N6520012 N6120009 N6120022
101680-794EA 4873.85 USD
101680-794 101680-790 101680-792
Complete Capillary Injector Add-On Kits, PerkinElmer
Chromatography Supplies Chromatography Injectors
Información de suministro: Kit includes split/splitless injector in module with heater and sensor, 0-60 psig pressure regulator, and transducer for pressure readout. For manual gas control.