Unconditioned Thermal Desorber Tubes, PerkinElmer
Supplier: Revvity Health Sciences
N9307052 N9307055
101689-402PK 1351.38 USD
101689-402 101689-406 101689-410 101689-398 101689-400 101689-408 101689-412
Unconditioned Thermal Desorber Tubes, PerkinElmer
Chromatography Supplies Chromatography Thermal Desorber Tubes
These tubes are offered with a variety of sorbent packing materials for many GC applications including indoor and outdoor air monitoring, analysis of avors and fragrances and the analysis of outgassing from packaging, polymers, pharmaceuticals and semi-conductor material.
Envase: These tubes are unconditioned and ship with plastic end caps for short-term storage.