Hematin ≥90.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Hydroxyhemin, Ferrihemate

H1156-250GMBL H1156-25GM H1156-5GM
75872-608EA 16165.65 USD
75872-608 75872-610 75872-612
Hematin ≥90.0%
Hematin

Hematin is a porphyrin that contains a ferric iron ion with a chloride ligand. It is typically used to treat porphyria attacks, primarily in acute intermittent porphyria. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


Hematin can be used to treat porphyria attacks, primarily in acute intermittent porphyria.


Synonym: Hydroxyhemin

Formula: C₃₄H₃₃FeN₄O₅
MW: 633.49 g/mol
Storage Temperature: Low Temperature Freezer (–40°C)
MDL Number: MFCD00011615
CAS Number: 15489-90-4

Order Now

Specification Test Results

Purity Min. 90.0%
Water Max. 3.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR