Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl) ≥99%

Supplier: CHEM IMPEX INTL INC MS

Synonyms: H-Gly-OtBu.HCl, Glycine tert-butyl ester hydrochloride, H-Gly-OtBu·HCl

03072-1KG 03072-1G 03072-25G 03072-100G 03072-5G 03072-250G
102554-624EA 2408.86 USD
102554-624 102574-934 102574-936 102589-846 102618-020 102638-782
Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl) ≥99%
Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl)
Formula: C₆H₁₄ClNO₂
MW: 167.64 g/mol
Melting Pt: 128…141 °C
MDL Number: MFCD00058255
CAS Number: 27532-96-3

Order Now

Specification Test Results

Water Content (Karl Fisher) ≤1%
Infrared Spectrum Conforms to Structure
Analysis by Mass Spectra Conforms to Standard
Loss on Drying ≤1% (70 ºC, 2h)
¹H NMR Conforms to Structure
Purity by HPLC ≥99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR