ESD Safe Pens, Transforming Technologies
Supplier: Transforming Technologies
DC2000R DC2000
76209-884PK 63.22 USD
76209-884 76210-050
ESD Safe Pens, Transforming Technologies
Pens Ink Pens
The DC2000 series static dissipative ballpoint pen is perfect for use in static controlled manufacturing facilities.
  • ESD safe pens are compatible with cleanrooms and other controlled areas
  • Printed with an ESD symbol in yellow
  • Fine point
Use this pen in ESD protected areas or any controlled area where static generation is a concern. The pens do not tribo-charge and are printed with an ESD symbol in yellow. Available in black, blue, or red ink.
Certificaciones: Meets or exceeds requirements of ANSI ESDS20.20.
Envase: Sold in packs of 10.