Perchloric acid 0.100 N (N/10) in glacial acetic acid for non-aqueous titration
Supplier: Ricca Chemical
|
Danger
|
R5515000-4C
R5515000-1C
R5515000-20F
R5515000-2.5D
R5515000-500C
RC55151EA
402
USD
RC55151
RCR5515000-1C
RCR5515000-20F
RCR55150002.5D
RCR5515000500C
Perchloric acid 0.100 N (N/10) in glacial acetic acid for non-aqueous titration
Perchloric acid
Formula:
HClO₄ MW: 100.46 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011325 CAS Number: 7601-90-3 UN: 2789 ADR: 8 (3),II |
Specification Test Results
Appearance | Colorless to light yellow liquid |
Assay (vs. Potassium Hydrogen Phthalate/Crystal Violet) | 0.0999-0.1001 N at 20°C |
Water (H₂O) | 0.020-0.050 % (w/w) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...