Oxalic acid dihydrate ACS
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Ethanedioic acid dihydrate
RDCO0200-125B1
RDCO0200-500B1
RCRDCO020125B1EA
116
USD
RCRDCO020125B1
RCRDCO020500B1
Oxalic acid dihydrate ACS
Oxalic acid dihydrate
Formula:
HO₂CCO₂H·2H₂O MW: 126.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149102 CAS Number: 6153-56-6 UN: 3261 ADR: 8,III |
Specification Test Results
Assay | 99.5-102.5 % |
Heavy Metals (as Pb) | max 5 ppm |
Nitrogen Compounds (as N) | max 0.001 % |
Sulfate (SO₄) | max 0.005 % |
Chloride (Cl) | max 0.002 % |
Calcium (Ca) | max 0.001 % |
Iron (Fe) | max 2 ppm |
Insoluble Matter | max 0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...