Molybdenum, disc, drilled apertures 2 mm O.D., Thickness: 0.24 mm

Supplier: Electron Microscopy Sciences

40010-MO 40020-MO 40030-MO 40050-MO 40100-MO 40150-MO
102092-542EA 421.77 USD
102092-542 102092-544 102092-546 102092-548 102092-550 102092-552
Molybdenum, disc, drilled apertures 2 mm O.D., Thickness: 0.24 mm
Molybdenum

Hole Sizes not listed are available upon special request.

Formula: Mo
MDL Number: MFCD00003465
CAS Number: 7439-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR