Molybdenum, disc, drilled apertures 6.35 mm O.D., Thickness: 0.14 mm
Supplier: Electron Microscopy Sciences
471000-MOS
47500-MOS
102092-698EA
224.52
USD
102092-698
102092-708
Molybdenum, disc, drilled apertures 6.35 mm O.D., Thickness: 0.14 mm
Molybdenum
Hole Sizes not listed are available upon special request.
Formula:
Mo MW: 95.94 g/mol Boiling Pt: 3617 Melting Pt: 2622 |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 Merck Index: 12,06317 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...