Molybdenum, disc, drilled apertures 4 mm O.D., Thickness: 0.165 mm

Supplier: Electron Microscopy Sciences

45010-MO 45020-MO 45030-MO 45050-MO 45070-MO 45100-MO
102092-672EA 352.38 USD
102092-672 102092-674 102092-676 102092-678 102092-680 102092-682
Molybdenum, disc, drilled apertures 4 mm O.D., Thickness: 0.165 mm
Molybdenum

Hole Sizes not listed are available upon special request.

Formula: Mo
MDL Number: MFCD00003465
CAS Number: 7439-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR