Molybdenum, disc, drilled apertures 3 mm O.D., Thickness: 0.24 mm
Supplier: Electron Microscopy Sciences
44005-MO
44010-MO
44015-MO
44020-MO
44025-MO
44030-MO
44040-MO
44050-MO
44060-MO
44070-MO
44100-MO
44150-MO
44200-MO
44300-MO
44400-MO
102092-642EA
505.15
USD
102092-642
102092-644
102092-646
102092-648
102092-650
102092-652
102092-654
102092-656
102092-658
102092-660
102092-662
102092-664
102092-666
102092-668
102092-670
Molybdenum, disc, drilled apertures 3 mm O.D., Thickness: 0.24 mm
Molybdenum
Hole Sizes not listed are available upon special request.
Formula:
Mo |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...