Molybdenum, sputtering target sputter coater cathodes

Supplier: Electron Microscopy Sciences

91006-MO 91007-MO 91008-MO 91009-MO 91010-MO 91013-MO 91014-MO 91015-MO 91016-MO 91017-MO 91018-MO 91019-MO 91020-MO
102099-408EA 146.06 USD
102099-408 102099-438 102099-468 102099-498 102099-526 102099-558 102099-588 102099-618 102099-648 102099-678 102099-708 102099-738 102099-768
Molybdenum, sputtering target sputter coater cathodes
Molybdenum

EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.


EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.

Formula: Mo
MDL Number: MFCD00003465
CAS Number: 7439-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR