Magid® TouchMaster® Lightweight Seamless Lisle Gloves, Magid
Supplier: Magid Glove
KTP1 KTP14
11310-592BG 21.97 USD
11310-592 11310-594
Magid® TouchMaster® Lightweight Seamless Lisle Gloves, Magid
Gloves
Machine-knit lisle glove that fits like a dress glove.
  • Color-coded sizes for easy sorting
  • Breathable lightweight fabric
This lightweight, seamless style has a reversible pattern for longer wear, and features a cotton and polyester blend for a cool and comfortable fit. These allso feature an over-edge hem to ensure a snug fit and prevent unraveling when washed. This TouchMaster glove is perfect for inspection or any general purpose use.