Magid® KnitMaster® Machine Knit Nylon Gloves, Magid
Supplier: Magid Glove
30NYMX 30NYSX 30NYLX
89134-378PK 16.45 USD
89134-378 89134-380 89134-382 89410-270 89410-272 89410-274
Magid® KnitMaster® Machine Knit Nylon Gloves, Magid
Gloves
The Magid KnitMaster gloves are highly useful, form fitting, 100% nylon seamless machine knit glove.
  • Lightweight 100% stretch nylon design
  • Seamless machine knit glove
  • Reversible pattern and knit wrists add longevity
  • Color-coded overedge to prevent unraveling
It features a reversible pattern for longer wear and a comfortable knit wrist for a secure fit. Excellent where lint is a problem, this glove has a high density knit for less snag and penetration.
These are great glove for paint departments and is also useful as a liner for the electronic and computer industry.