Ruthenium 5% on activated carbon powder wetted with water 27 µm for use as a catalyst, reduced , BASF Catalyst
Supplier: SPECTRUM CHEMICAL MFG CORP
SP-400-100G
SP-400-25G
SP-400-5G
76124-516EA
749.65
USD
76124-516
76124-518
76124-520
Ruthenium 5% on activated carbon powder wetted with water 27 µm for use as a catalyst, reduced , BASF Catalyst
Ruthenium
5 Percent Ruthenium on Activated Carbon Powder, Reduced, Water Wet, 27 microns, BASF Catalyst
Formula:
Ru Storage Temperature: Ambient |
MDL Number:
MFCD00011207 CAS Number: 7440-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...