Rhodium 5% on alumina powder, dry 70 µm for use as a catalyst, reduced , BASF Catalyst

Supplier: SPECTRUM CHEMICAL MFG CORP

SP-301-10G SP-301-2G SP-301-50G
76169-178EA 982.4 USD
76169-178 76169-180 76169-182
Rhodium 5% on alumina powder, dry 70 µm for use as a catalyst, reduced , BASF Catalyst
Rhodium

5 Percent Rhodium on Alumina Powder, Reduced, Dry, 70 microns, BASF Catalyst

Formula: Rh
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS Number: 7440-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR