Rhodium 5% on activated carbon powder wetted with water 27 µm for use as a catalyst, reduced , BASF Catalyst
Supplier: SPECTRUM CHEMICAL MFG CORP
SP-300-10G
SP-300-1G
SP-300-50G
SP-300-5G
76124-508EA
488
USD
76124-508
76124-510
76124-512
76124-514
Rhodium 5% on activated carbon powder wetted with water 27 µm for use as a catalyst, reduced , BASF Catalyst
Rhodium
5 Percent Rhodium on Activated Carbon Powder, Reduced, Water Wet, 27 microns, BASF Catalyst
Formula:
Rh MW: 102.9 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011201 CAS Number: 7440-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...