Platinum 5% on alumina powder, dry 70 µm for use as a catalyst, reduced , BASF Catalyst
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Platinium
SP-204-100G
SP-204-25G
SP-204-5G
76124-496EA
1727.55
USD
76124-496
76124-498
76124-500
Platinum 5% on alumina powder, dry 70 µm for use as a catalyst, reduced , BASF Catalyst
Platinum
5 Percent Platinum on Alumina Powder, Reduced, Dry, 70 microns, BASF Catalyst
Formula:
Pt MW: 195.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...