Airfree® Schlenk Connecting Adapters, Chemglass
Supplier: Chemglass
AF-0076-01 AF-0076-02
80064-268EA 322.58 USD
80064-268 80064-270
Airfree® Schlenk Connecting Adapters, Chemglass
Adapters Adapters, Joints with Tube Connectors
Adapter has two #15 O-Ring joints connected in-line via a Chem-Cap® valve.
  • Available with either 0 to 4 mm or 0 to 8 mm valve sizes
Información de suministro: Supplied complete with two #116 Viton O-Rings.