Airfree® Schlenk Adapters, Vacuum or Inert Gas, with Stopcock, Chemglass
Supplier: Chemglass
AF-0509-B-01 AF-0509-B-02 AF-0509-B-03 AF-0509-B-04
80064-546EA 254.84 USD
80064-546 80064-548 80064-550 80064-552
Airfree® Schlenk Adapters, Vacuum or Inert Gas, with Stopcock, Chemglass
Adapters Adapters, Conical Ground Joints
Adapter has a T-Bore stopcock permitting vessels with a standard taper outer joint to be purged and evacuated.
  • Stopcocks have 9 mm O.D. medium wall arms
  • T-bore stopcock
Información de suministro: Available with a choice of a 4 mm glass high vacuum stopcock with a stainless steel constant pressure retention device or 4 mm PTFE stopcock.