Airfree® Schlenk Thermocouple Gauge Adapter, Chemglass
Supplier: Chemglass
AF-0306-03 AF-0306-10
89028-174EA 436.3 USD
89028-174 89028-176
Airfree® Schlenk Thermocouple Gauge Adapter, Chemglass
Adapters Adapters, Joints with Tube Connectors
A (0 to 12 mm) Chem-Vac™ Chem-Cap® valve with an O-Ring joint for connection to the right-hand side of (80064-398) vacuum manifold.
  • Available for #30 O-Ring joints
Información de suministro: Complete gauge consists of a glass portion, a stainless steel bushing drilled and tapped for 1/8" NPT, permitting direct connection to gauge tube, and an open top GL-25 screw cap.