Anti-APP Mouse Monoclonal Antibody [clone: WO2]

Supplier: Enzo Life Sciences
ABS386-0100
76284-702EA 670 USD
76284-702
Anti-APP Mouse Monoclonal Antibody [clone: WO2]
Antibodies
Recombinant monclonal antibody (Mouse IgG2aκ).

Recommended Applications: ELISA, IHC (PS), WB

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: WO2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2aκ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR