Airfree® Schlenk Adapter, Chemglass
Supplier: Chemglass
AF-0501-01 AF-0501-02
80064-478EA 215.19 USD
80064-478 80064-480
Airfree® Schlenk Adapter, Chemglass
Adapters Adapters, Conical Ground Joints
Adapter allows standard single-neck round bottom flasks to be used as storage vessels..
  • Equipped with 0 to 4 mm Chem-Vac™ Chem-Cap® plug
Sampling can be accomplished by removing the 0 to 4 mm Chem-Vac™ Chem-Cap® plug, while maintaining inert gas purge through sidearm.
Información para pedidos: Available with a choice of 14/20 or 24/40 standard taper inner joint.