1,2-Epoxybutane 99%

Supplier: Thermo Scientific Chemicals
Danger

H36581-14 H36581-22
AAH36581-14EA 26 USD
AAH36581-14 AAH36581-22
1,2-Epoxybutane 99%
1,2-Epoxybutane
Formula: C₄H₈O
Storage Temperature: Ambient
MDL Number: MFCD00005153
CAS Number: 106-88-7
EINECS: 203-438-2
UN: 3022
ADR: "3",II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR