Anti-SCF Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P71BT-25UG 500-P71BT-50UG
10775-624EA 339.8 USD
10775-624 10775-626 10775-628
Anti-SCF Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine SCF. Anti­Murine SCF­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: SCF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR