Anti-RELMbeta Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P217BT-25UG 500-P217BT-50UG
10776-972EA 339.8 USD
10776-972 10776-974 10776-976
Anti-RELMbeta Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human RELMβ. Anti­Human RELMβ­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: RELMbeta
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen Symbol RELMbeta
Antigen Name RELMbeta
Environmentally Preferable
Antibody Type Secondary
Clonality Polyclonal
Conjugation Biotin
Reactivity Human
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
Immunogen Recombinant Human RELMβ
Form Lyophilized
Molecular Weight 19.0 kDa
Purification Recombinant Human RELMβ
Storage Buffer See Product Data Sheet for more information.
Storage Temperature Store at –20 °C prior to reconstitution.
Shipping Temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR