Anti-BD-1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P253BT-50UG 500-P253BT-25UG
10771-496EA 434.63 USD
10771-496 10771-498 10777-296
Anti-BD-1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human BD-1. Anti­Human BD-1­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: BD-1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR