Anti-EGF Receptor (EGFR) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P306BT-25UG 500-P306BT-50UG
10781-634EA 354.54 USD
10781-634 10781-636 10781-638
Anti-EGF Receptor (EGFR) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human EGF Receptor (EGFR). Anti­Human EGF Receptor (EGFR)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: EGF Receptor (EGFR)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR