Anti-KC (CXCL1) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P115-50UG 500-P115-100UG 500-P115-1MG
10776-070EA 354.54 USD
10776-070 10776-072 10776-074
Anti-KC (CXCL1) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine KC (CXCL1). Anti­Murine KC (CXCL1)­specific antibody was purified by affinity chromatography employing an immobilized Murine KC matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: KC (CXCL1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR