Anti-GM-CSF Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P65-50UG 500-P65-100UG 500-P65-1MG
10775-558EA 339.8 USD
10775-558 10775-560 10775-562
Anti-GM-CSF Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine GM-CSF. Anti­Murine GM-CSF­specific antibody was purified by affinity chromatography employing an immobilized Murine GM-CSF matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: GM-CSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR