Anti-FGF-9 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P66-50UG 500-P66-100UG 500-P66-1MG
10775-570EA 339.8 USD
10775-570 10775-572 10775-574
Anti-FGF-9 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine FGF-9. Anti­Murine FGF-9­specific antibody was purified by affinity chromatography employing an immobilized Murine FGF-9 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: FGF-9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR