Anti-PF-4 (CXCL4) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P05-50UG 500-P05-100UG 500-P05-1MG
10774-994EA 339.8 USD
10774-994 10774-996 10774-998
Anti-PF-4 (CXCL4) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human PF-4 (CXCL4). Anti­Human PF-4 (CXCL4)­specific antibody was purified by affinity chromatography employing an immobilized Human PF-4 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: PF-4 (CXCL4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR