Anti-NOV Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P257-50UG 500-P257-100UG 500-P257-1MG
10777-322EA 339.8 USD
10777-322 10777-324 10777-326
Anti-NOV Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human NOV. Anti­Human NOV­specific antibody was purified by affinity chromatography employing an immobilized Human NOV matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: NOV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR