Anti-NNT-1/BCSF-3 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P186-50UG 500-P186-100UG 500-P186-1MG
10771-476EA 339.8 USD
10771-476 10771-478 10771-480
Anti-NNT-1/BCSF-3 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human NNT-1/BCSF-3. Anti­Human NNT-1/BCSF-3­specific antibody was purified by affinity chromatography employing an immobilized Human NNT-1/BCSF-3 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: NNT-1/BCSF-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR